missing translation for 'onlineSavingsMsg'
Learn More
Learn More
u-Plasminogen Activator/Urokinase Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-2419
This item is not returnable.
View return policy
Description
u-Plasminogen Activator/Urokinase Polyclonal antibody specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Specifications
| u-Plasminogen Activator/Urokinase | |
| Polyclonal | |
| Unconjugated | |
| Tris (pH 7.3), 0.5% BSA | |
| ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator | |
| Peptide with sequence C-HFLPWIRSHTKEEN corresponding to C-Terminus according to NP_002649.1, NP_001138503.1. | |
| 0.1 mg | |
| Cancer | |
| 5328 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 8 μg/mL, Immunohistochemistry-Paraffin 8 μg/mL, Peptide ELISA Detection limit 1:6000 | |
| NP_002649.1 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction