missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TYKi Antibody (CL14486), Novus Biologicals™
Mouse Monoclonal Antibody
433.00€ - 629.00€
Specifications
| Antigen | TYKi |
|---|---|
| Clone | CL14486 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043550
|
Novus Biologicals™
NBP3-26858-100ul |
100 μL |
629.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043214
|
Novus Biologicals™
NBP3-26858-25ul |
25 μL |
433.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TYKi Monoclonal antibody specifically detects TYKi in Human samples. It is validated for Western BlotSpecifications
| TYKi | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| PBS, pH 7.2, 40% glycerol | |
| 129607 | |
| IgG2b | |
| Protein A purified |
| CL14486 | |
| Western Blot 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, cytidylate kinase 2, EC 2.7.4.14, mitochondrial, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member, TMPK2, TYKi, UMP-CMPK2 | |
| This antibody was developed against a Synthetic Peptide of Q5EBM0. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title