missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYKi Antibody (CL14486) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26858MFV500
This item is not returnable.
View return policy
Description
TYKi Monoclonal antibody specifically detects TYKi in Human samples. It is validated for Western Blot
Specifications
| TYKi | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, cytidylate kinase 2, EC 2.7.4.14, mitochondrial, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member, TMPK2, TYKi, UMP-CMPK2 | |
| This antibody was developed using a synthetic peptide derived from Q5EBM0, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Protein Kinase | |
| 129607 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot | |
| CL14486 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction