Learn More
IKA™ MIXSTER Sets
GREENER_CHOICE
Product Code. 17705878 Shop All IKA Products
Click to view available options
Includes:
2 x Stirring Unit, 50 Stir Bars
3 x Stirring Unit, 75 Stir Bars
4 x Stirring Unit, 100 Stir Bars
Stirring Unit, 25 Stir Bars
Unit Size:
Each
Pack of 2
Pack of 3
Pack of 4
Product Code. 17705878
Brand: IKA™ 0020027389
MIXSTER is a universal and compact mixing device with magnetic drive.
In the default version, MIXSTER is a powerful single-position magnetic stirrer for mixing up to 5 L at a maximum speed of 3000 rpm and a viscosity of up to 500 mPa. By connecting additional MIXSTER stirring units (available as accessories), MIXSTER can be expanded to a multi-position magnetic stirrer with up to 30 stirring positions. Stirring positions can be controlled either separately or simultaneously (leader-follower principle).
  • With multi-position stirring attachment TW.IX, the drive is transformed into a magnetic stirrer with 9 stirring positions for efficient mixing of sample vessels with small volumes (8 to 60 mL)
  • TW.VX shaker attachment transforms universal magnetic drive into a vortex or orbital shaker
  • MIXSTER enables all information to be conveyed without any alpha-numeric display via its intuitive and chemically resistant 360° LED glass display
  • During operation, speed or power required for stirring can be displayed
  • Viscosity changes can be observed and displayed as a percentage
  • Mixing can be started by pressing a button thanks to integrated beaker recognition by placing or touching the beaker
  • Reverse function changes the direction of rotation at definable time intervals, leading to a significant improvement of the mixing result, especially with samples that are difficult to mix
  • High protection class IP 66 allows operation under harsh conditions
  • MIXSTER can be cleaned under the tap, for example, or operated in an incubator at temperatures up to 40°C
  • Compact and innovative design has already been awarded the Reddot Award 2022
  • Particularly suitable for use in laboratories with limited space
  • Depending on number of devices, a different power supply may be needed (included power supply is designed to power up to two devices)
TRUSTED_SUSTAINABILITY
Specifications
Speed 100 to 3000 rpm
Stirring Positions 3
Top Plate Material Glass
Capacity 5 L
Display LED
Shape Rectangle
Voltage 24 V
Wattage 10 W
Height (Metric) 24 mm
Length (Metric) 135 mm
Top Plate Diameter (Metric) 124 mm
Width (Metric) 135 mm
Amperage 400 mA
Frequency 50/60 Hz
Plug Type Universal
Certifications/Compliance CE
Warranty and Service Offering 3 Year
Color Black
Includes 3 x Stirring Unit, 75 Stir Bars
Material Aluminum
Protection Class IP 66
Speed Control Touch Key
Timer Yes
Type Mixing Device
Shipping Weight (Metric) 1.4 kg