missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TWIST2 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149165
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Binds to the E-box consensus sequence 5'-CANNTG-3' as a heterodimer and inhibits transcriptional activation by MYOD1, MYOG, MEF2A and MEF2C. Also represses expression of proinflammatory cytokines such as TNFA and IL1B. Involved in postnatal glycogen storage and energy metabolism. Inhibits the premature or ectopic differentiation of preosteoblast cells during osteogenesis, possibly by changing the internal signal transduction response of osteoblasts to external growth factors.
Specifications
| TWIST2 | |
| Polyclonal | |
| Unconjugated | |
| TWIST2 | |
| AMS; BBRSAY; bHLHa39; Class A basic helix-loop-helix protein 39; dermis expressed 1; dermis-expressed protein 1; DERMO1; Dermo-1; dermo-1 protein; FFDD3; SETLSS; twist basic helix-loop-helix transcription factor 2; twist family bHLH transcription factor 2; twist homolog 2; TWIST2; twist-related bHLH protein Dermo1; twist-related protein 2 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 117581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8WVJ9 | |
| TWIST2 | |
| E. coli-derived recombinant human Twist-2, Met1-Ser159. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction