missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Twist-1 Antibody (10E4E6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
232.00€ - 639.00€
Specifications
| Antigen | Twist-1 |
|---|---|
| Clone | 10E4E6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Flow (Intracellular), CyTOF-ready, Knockdow |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18091704
|
Novus Biologicals
NBP2-37364SS |
0.025 mL |
232.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18748543
|
Novus Biologicals
NBP2-37364 |
0.1 mL |
639.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Twist-1 Monoclonal specifically detects Twist-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready, Knockdown Validated.Specifications
| Twist-1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS with 0.05% Sodium Azide | |
| 7291 | |
| Purified recombinant fragment of human TWIST1 (AA: 9-74) expressed in E. Coli. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 10E4E6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Flow (Intracellular), CyTOF-ready, Knockdow | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, twist-related protein 1 | |
| TWIST1 | |
| IgG1 | |
| Protein A or G purified | |
| 21 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title