Learn More
Eppendorf™ twin.tec™ Trace PCR Plates - PCR Plates
Shop All Eppendorf Products
Click to view available options
Color:
Colorless
Crystal Blue
Fuchsia
Product Type:
Trace PCR Plate
Trace PCR Plate LoBind™
Unit Size:
Pack of 25
Description
- Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
- Unique laser-engraved optical guiding grid
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Spezifikation
Spezifikation
| Barcode | No Barcode |
| Format | Standard |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Packaging | 5 Bags x 5 Plates |
| Product Line | Eppendorf |
| Color | Fuchsia |
| Product Type | Trace PCR Plate |
| Skirt Style | Skirted |
| Material | Polypropylene, Polycarbonate |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur