missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWF2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 470.00€
Specifications
| Antigen | TWF2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642922
|
Novus Biologicals
NBP2-94467-0.02ml |
0.02 mL |
188.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660512
|
Novus Biologicals
NBP2-94467-0.1ml |
0.1 mL |
470.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TWF2 Polyclonal antibody specifically detects TWF2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| TWF2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 11344 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| A6-related protein, A6RPFLJ56277, hA6RP, Protein tyrosine kinase 9-like, protein tyrosine kinase 9-like (A6-related protein), PTK9L protein tyrosine kinase 9-like (A6-related protein), PTK9LA6r, twinfilin, actin-binding protein, homolog 2 (Drosophila), Twinfilin-1-like protein, twinfilin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-349 of human TWF2 (NP_009215.1). MAHQTGIHATEELKEFFAKARAGSVRLIKVVIEDEQLVLGASQEPVGRWDQDYDRAVLPLLDAQQPCYLLYRLDSQNAQGFEWLFLAWSPDNSPVRLKMLYAATRATVKKEFGGGHIKDELFGTVKDDLSFAGYQKHLSSCAAPAPLTSAERELQQIRINEVKTEISVESKHQTLQGLAFPLQPEAQRALQQLKQKMVNYIQMKLDLERETIELVHTEPTDVAQLPSRVPRDAARYHFFLYKHTHEGDPLESVVFIYSMPGYKCSIKERMLYSSCKSRLLDSVEQDFHLEIAKKIEIGDGAELTAEFLYDEVHPKQHAFKQAFAKPKGPGGKRGHKRLIRGPGENGDDS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title