missing translation for 'onlineSavingsMsg'
Learn More

Tween™80, ∽75% oleic acid, MP Biomedicals™

Product Code. p-4404570 Shop All MP Biomedical Products
Click to view available options
Quantity:
0.5 L
1 L
4 L
Unit Size:
1 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 11483601

Brand: MP Biomedicals 0210317090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-70-3
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
Molecular Weight (g/mol) NaN
MDL Number MFCD01779641
InChI Key ZGEQUSWSYZZNAI-CLFAGFIQNA-N
Synonym Polysorbate 80, Polyoxyethylene sorbitan monooleate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Liquid
Density 1.08g/mL
Quantity 0.5 L
Assay Percent Range ∽75%
Chemical Name or Material Tween 80
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.