CAS RN 9005-70-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
(C2H4O)D(C2H4O)C(C2H4O)B(C2H4O)AC60H107O8
InChi Key:
—
SMILES:
CCCCCCCC\C=C/CCCCCCCC(-*)=O.CCCCCCCC\C=C/CCCCCCCC(-*)=O.CCCCCCCC\C=C/CCCCCCCC(-*)=O.[H][C@]1(CO[C@]([H])([C@@H](COCCO-*)OCCO-*)[C@@]1([H])OCCO-*)OCCO-*
1
–
2
of
2
results
1
Polysorbate 85
CAS: 9005-70-3 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7 Molecular Weight (g/mol): NaN MDL Number: MFCD01779641 InChI Key: ZGEQUSWSYZZNAI-CLFAGFIQNA-N Synonym: Polyoxyethylene (20) Sorbitan Trioleate,Polysorbate 85 SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
2
Tween™80, ∽75% oleic acid, MP Biomedicals™
CAS: 9005-70-3 Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7 Molecular Weight (g/mol): NaN MDL Number: MFCD01779641 InChI Key: ZGEQUSWSYZZNAI-CLFAGFIQNA-N Synonym: Polysorbate 80,Polyoxyethylene sorbitan monooleate SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO