Learn More
Tween™80, ∽75% oleic acid, MP Biomedicals™
Product Code. 11451981 Shop All MP Biomedical Products
Click to view available options
Quantity:
0.5 L
1 L
4 L
Unit Size:
1 L
4 L
500mL
Product Code. 11451981
Brand: MP Biomedicals 0219472504
Chemical Identifiers
CAS 9005-70-3
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
Molecular Weight (g/mol) NaN
MDL Number MFCD01779641
InChI Key ZGEQUSWSYZZNAI-CLFAGFIQNA-N
Synonym Polysorbate 80, Polyoxyethylene sorbitan monooleate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
Specifications
Color Yellow
Physical Form Liquid
Density 1.08g/mL
Quantity 4 L
Assay Percent Range ∽75%
Chemical Name or Material Tween 80