missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUG Antibody (4A11A6G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
168.00€
Specifications
| Antigen | TUG |
|---|---|
| Clone | 4A11A6G11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1: 500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot |
Description
TUG Monoclonal specifically detects TUG in Mouse samples. It is validated for Western Blot, ELISA.Specifications
| TUG | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, alveolar soft part sarcoma chromosome region, candidate 1, Alveolar soft part sarcoma locus, ASPLUBX domain-containing protein 9, ASPSrenal cell carcinoma gene on chromosome 17, RCC17renal cell carcinoma, papillary, 17, Renal papillary cell carcinoma protein 17, tether containing UBX domain for GLUT4, TUG, UBX domain protein 9, UBXD9ASPCR1, UBXN9FLJ45380 | |
| ASPSCR1 | |
| IgG1 | |
| Affinity Purified |
| 4A11A6G11 | |
| Western Blot 1: 500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Mouse | |
| 79058 | |
| Purified recombinant fragment of TUG expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title