Learn More
TUFM Antibody (CL2245) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36753MFV500
Product Code. 30296159
  • 759.00€ / 0.10mL
Description
TUFM Monoclonal antibody specifically detects TUFM in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TUFM
Monoclonal
mFluor Violet 500 SE
EFTu, EF-Tu, EFTUCOXPD4, EF-TuMT, elongation factor Tu, mitochondrial, P43, Tu translation elongation factor, mitochondrial
This antibody was developed using a recombinant protein derived from P49411, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL2245
50mM Sodium Borate
Mouse
Protein A purified
RUO
7284
Store at 4⪚C in the dark.