Learn More
SPEX Certiprep™ Tube Holder
Product Code. 15320462 Shop All SPEX Certiprep Products
Product Code. 15320462
Brand: SPEX Certiprep™ 1686SSP
This item requires a price update prior to being ordered.
Request a Price for this item here.
Thick, sturdy foam block that holds 6 standard 50ml tubes in the MiniG homogeniser from SPEX SamplePrep.
  • Rugged foam block to hold sample tubes securely in 1690 adjustable clamp for the 1600 MiniG.
  • Process 6 samples at one time.
Specifications
Capacity (Metric) 50 mL
For Use With (Equipment) 1690 adjustable clamp for the 1600 MiniG
Holds 6 Tubes
Material Foam Block
Packaging Quantity 6 Pk.