Learn More
Cytiva Tube Holder
Product Code. p-8001357 Shop All Cytiva Products
Click to view available options
For Use With:
95 Tubes
Frac-900
Unit Size:
1 set
Pack of 2
Product Code. 12554307
Brand: Cytiva 18112468
Tube Holder for Frac-900 or Frac-901
The tube guide can be placed at different heights to accommodate different length of tubes.
TRUSTED_SUSTAINABILITY
Specifications
Type Tube Holder
Packaging Quantity 2 Pack
Includes Tube Holder and Guide
For Use With Frac-900