missing translation for 'onlineSavingsMsg'
Learn More

TTL Antibody (OTI3G5), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18114667 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18114667 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18114667 Supplier Novus Biologicals Supplier No. NBP274668AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TTL Monoclonal specifically detects TTL in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TTL
Applications Western Blot
Classification Monoclonal
Clone OTI3G5
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 6.3.2.25,2410003M22Rik, MGC46235, tubulin tyrosine ligase, tubulin-tyrosine ligase, tubulin--tyrosine ligase
Gene Symbols TTL
Host Species Mouse
Immunogen Full length recombinant protein of human TTL (NP_714923) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 150465
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.