Learn More
TTF-1/ NKX2-1 Antibody (CL14528) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26861AF750
Product Code. 30304304
  • 770.00€ / 0.10mL
Description
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TTF-1 / NKX2-1
Monoclonal
Alexa Fluor 750
benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2
This antibody was developed using a synthetic peptide derived from P43699, with the exact immunogen sequence remaining proprietary.
0.1 mL
Membrane Trafficking and Chaperones, Tumor Biomarkers
7080
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot
CL14528
50mM Sodium Borate
Mouse
Protein A purified