missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF-1/ NKX2-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€ - 507.00€
Specifications
| Antigen | TTF-1 / NKX2-1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600459
|
Novus Biologicals
NBP2-41160-0.025mg |
0.025 mg |
168.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18789283
|
Novus Biologicals
NBP2-41160 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TTF-1 / NKX2-1 Polyclonal specifically detects TTF-1 / NKX2-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TTF-1 / NKX2-1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 | |
| NKX2-1 | |
| IgG | |
| Affinity Purified | |
| 41, 45 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Tumor Biomarkers | |
| PBS with 0.02% Sodium Azide | |
| 7080 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human NKX2-1. The immunogen is located within the first 50 amino acids of NKX2-1. Amino Acid Squence: LSPLEESYKKVGMEGG | |
| Primary | |
| NKX2-1 antibody is predicted to not cross-react with other NK2 homeobox family members. At least three isoforms of NKX2-1 are known to exist; this antibody will detect all three. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title