missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC30A Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68232-1-IG-20UL
This item is not returnable.
View return policy
Description
The TTC30A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TTC30A. This antibody recognizes human, mouse, rat antigen. The TTC30A antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| TTC30A | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ13946; FLJ77601 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 72-76 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1A11B3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 92104 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction