missing translation for 'onlineSavingsMsg'
Learn More

TSSK4 Antibody, Novus Biologicals™

Product Code. 18129571 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18129571 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18129571 Supplier Novus Biologicals Supplier No. NBP220751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TSSK4 Polyclonal specifically detects TSSK4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSSK4
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q6SA08
Gene Alias C14orf20, EC 2.7.11, EC 2.7.11.1, MGC133264, serine/threonine kinase 22E, Serine/threonine-protein kinase 22E, STK22E, Testis-specific kinase 4, testis-specific serine kinase 4, testis-specific serine/threonine-protein kinase 4, TSK-4, TSSK-4, TSSK5TSK4
Gene Symbols TSSK4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TSSK4. The exact sequence is proprietary.
Molecular Weight of Antigen 37 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 283629
Test Specificity Human. Species reactivity based on sequence identity: Mouse (85%), Bovine (89%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.