missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSPAN9 Monoclonal Antibody (590409)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561689
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
TSPAN9 includes the tetraspan family protein members which are characterized by four predicted transmembrane domains, and are thought to be involved in physiological processes such as tissue differentiation, immunological responses, and sperm-egg fusion. TSPAN9 has recently been identified as a platelet tetraspanin and a component of tetraspanin microdomains that include the collagen receptor GPVI (glycoprotein VI) and integrin alpha6beta1 but not the von Willebrand receptor GPIbalpha or the integrins alphaIIbbeta3 or alpha2beta1, suggesting that TSPAN9 may act to regulate platelet function in concert with other tetraspanins and their associated proteins.
Specifications
| TSPAN9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O75954 | |
| TSPAN9 | |
| HEK293 human embryonic kidney cell line transfected with human TSPAN9. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 590409 | |
| Unconjugated | |
| TSPAN9 | |
| 6720474K14Rik; 9430079M16Rik; AU018597; NET5; NET-5; new EST tetraspan 5; PP1057; RGD1304740; tetraspan NET-5; tetraspanin 9; tetraspanin-9; transmembrane 4 superfamily member tetraspan NET-5; Tspan9; tspan-9 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10867 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto