missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN7/TM4SF2 Antibody (CL0262) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52894MFV500
This item is not returnable.
View return policy
Description
TSPAN7/TM4SF2 Monoclonal antibody specifically detects TSPAN7/TM4SF2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TSPAN7/TM4SF2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| A15CCG-B7, CD231, CD231 antigen, Cell surface glycoprotein A15, Membrane component chromosome X surface marker 1, mental retardation, X-linked 58, MRX58, MXS1T-cell acute lymphoblastic leukemia associated antigen 1, T-cell acute lymphoblastic leukemia-associated antigen 1, tetraspanin 7, tetraspanin-7, TM4SF2tetraspanin protein, transmembrane 4 superfamily 2b, Transmembrane 4 superfamily member 2X chromosome, surface marker 1, transmembrane protein A15, Tspan-7 | |
| This antibody was developed using a recombinant protein derived from P41732, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| metabolism | |
| 7102 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0262 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction