missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37885-20ul
This item is not returnable.
View return policy
Description
TSPAN18 Polyclonal antibody specifically detects TSPAN18 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TSPAN18 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| tetraspanin 18, TSPAN | |
| A synthetic peptide corresponding to a sequence within amino acids 149-248 of mouse TSPAN18 (NP_899003.1).,, Sequence:, GVNGPEDFKLASVFRLLTLDTEEVPKACCRREPQTRDGVVLSREECQLGRNPFINKQGCYTVILNTFETYVYLAGAFAIGVLAIELFLMVFAMCLFRGIQ | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90139 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction