missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TSLP Antibody (tezepelumab), Novus Biologicals™

Human Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-28453-100ug

 View more versions of this product

Product Code. 30045942

  • 326.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TSLP Monoclonal antibody specifically detects TSLP in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSLP
Monoclonal
Unconjugated
Q969D9
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG2
Flow Cytometry, ELISA, Functional Assay
tezepelumab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
thymic stromal lymphopoietin
TSLP
100 μg
Biologically Active Proteins, Innate Immunity
85480
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.