missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHZ1 Monoclonal Antibody (OTI2G9), TrueMAB™, OriGene
Description
TSHZ1 Monoclonal antibody specifically detects TSHZ1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TSHZ1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI2G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | STC1 |
| Gene Accession No. | P52823 |
| Gene Alias | 5730407I04Rik; antigen NY-CO-33; CAA; D18Bwg1409e; Kiaa4206; mKIAA4206; Mtsh1; NY-CO-33; SDCCAG33; Serologically defined colon cancer antigen 3 homolog; serologically defined colon cancer antigen 33; Teashirt homolog 1; teashirt zinc finger family member 1; teashirt zinc finger homeobox 1; teashirt1; TSH1; Tshz1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?