missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSH Receptor Recombinant Human Monoclonal Antibody (11A5)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560830
This item is not returnable.
View return policy
Description
TSH Receptor Recombinant Monoclonal Antibody for Flow, ELISA
TSH Receptor gene is a membrane protein and a major controller of thyroid cell metabolism. The encoded protein is a receptor for thyrothropin and thyrostimulin, and its activity is mediated by adenylate cyclase. Defects in this gene are a cause of several types of hyperthyroidism. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| TSH Receptor | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P16473 | |
| TSHR | |
| Recombinant Human TSHR protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry | |
| 11A5 | |
| Unconjugated | |
| TSHR | |
| AI481368; CHNG1; hTSHR-I; hypothroid; hyt; LGR3; MGC141920; pet; seven transmembrane helix receptor; thyroid stimulating hormone receptor; thyroid stimulating hormone receptor, isoform 2; thyroid-stimulating hormone receptor; thyroid-stimulating hormone receptor variant; thyrotropin receptor; thyrotropin receptor-I, hTSHR-I; TSHR; TSH-R; TSHR; seven transmembrane domain G protein-coupled receptor; TSHRA | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 7253 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction