missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Antibody (TSH-116), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
531.00€
Specifications
| Antigen | TSH beta |
|---|---|
| Clone | TSH-116 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Janelia Fluor 549 |
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for ELISA, Radioimmune AssaysSpecifications
| TSH beta | |
| ELISA | |
| Janelia Fluor 549 | |
| Mouse | |
| Cancer, Signal Transduction | |
| 50mM Sodium Borate | |
| 7252 | |
| IgG2a | |
| Protein A purified |
| TSH-116 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Human thyrotropin. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title