Learn More
TSC22D4 Monoclonal Antibody (OTI3G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811733S
Encompass_Preferred
 View more versions of this product
Product Code. 17525944
  • 242.00€ / 30µL
Description
TSC22D4 Monoclonal antibody specifically detects TSC22D4 in Human samples. It is validated for Western Blot
Specifications
Specifications
TSC22D4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P00742
Mouse
Affinity chromatography
RUO
2159
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3G6
Unconjugated
F10
0610009M14Rik; 1700023B23Rik; AI415410; LOW QUALITY PROTEIN: TSC22 domain family protein 4; THG1; THG-1; Thg1pit; Thg-1pit; Tilz2; TSC22 domain family 4; TSC22 domain family member 4; TSC22 domain family protein 4; TSC22 domain family, member 4; Tsc22d4; tsc-22-like protein THG-1; TSC22-related inducible leucine zipper 2; TSC22-related-inducible leucine zipper protein 2
Human recombinant protein fragment corresponding to amino acids 41-488 of human F10 produced in E.coli.
30 μL
Primary
Human
Liquid
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.