missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76619-0.025mg
This item is not returnable.
View return policy
Description
TSC2 Polyclonal specifically detects TSC2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.
Specifications
| TSC2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7249 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2-4 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
| NP_000539 | |
| TSC2 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human TSC2. The immunogen is located within the first 50 amino acids of TSC2. Amino Acid Squence: TPRPNPRSAEGKQT | |
| 0.025 mg | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction