missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-200AF532
This item is not returnable.
View return policy
Description
TSC2 Polyclonal antibody specifically detects TSC2 in Human samples. It is validated for Western Blot
Specifications
| TSC2 | |
| Polyclonal | |
| Western Blot | |
| LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Tuberous Sclerosis 2 using the numbering given in Swiss-Prot entry P49815 (GeneID 7249). | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Hypoxia, Inhibitors Activators and Regulators, mTOR Pathway, Phospho Specific, Signal Transduction, Tumor Suppressors | |
| 7249 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction