missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase alpha/beta 1 Antibody (rTPSAB1/1963), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23713-20ug
This item is not returnable.
View return policy
Description
Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Tryptase alpha/beta 1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III | |
| Recombinant human Tryptase alpha/beta 1 protein fragment (around aa 100-250) (exact sequence is proprietary) | |
| 20 μg | |
| Immunology, metabolism | |
| 7177 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rTPSAB1/1963 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction