Learn More
Novus Biologicals™ Tryptase alpha/beta 1 Antibody (Genentech patent anti-Tryptase Beta 1) - Humanized, IgG4SP, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28370-100ug
 View more versions of this product
Product Code. 30045650
  • 338.00€ / 100µg
Description
Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tryptase alpha/beta 1
Monoclonal
Unconjugated
P20231
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG4
Flow Cytometry, ELISA, Functional Assay
Genentech patent anti-Tryptase Beta 1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III
TPSAB1
100 μg
Immunology, metabolism
7177