missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Trypsin Inhbitor (Ovomucoid) (Egg White)
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
#REF!
Specifications
Specifications
| For Use With (Application) | Immunoassay |
| Molecular Weight (g/mol) | ∽28kDa |
| Name | Trypsin Inhibitor (Ovomucoid) (Egg White) |
| Preparation Method | Mucoprotein and antitryptic factor purified from egg white described by Lineweaver and Murray, J. Biol. Chem., 171, 565 (1947) |
| Quantity | 1 g |
| Storage Requirements | Store at 4°C. Reconstituted with 0.001 N HCl at a concentration of 1.0 mg/ml. |
| Regulatory Status | RUO |
| Common Name | TRYPSIN |
| Biological Activity | 1mg inhibits at least 1.2 mg trypsin (Cat. # P5255). |
| Species | Chicken |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction