Learn More
TRX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13089-1-AP-150UL
 View more versions of this product
Product Code. 16871134
  • 410.00€ / 150µL
Description
Thioredoxins (Trx) are small, multi-functional proteins with oxidoreductase activity and are ubiquitous in essentially all living cells. Trx contains a redox-active disulfide/dithiol group within the conserved Cys-Gly-Pro-Cys active site. The two cystein residues in the conserved active centers can be oxidized to form intramolecular disulfide bonds. Reduction of the active site disulfide in oxidized Trx is catalyzed by Trx reductase with NADPH as the electron donor. The reduced Trx is a hydrogen donor for ribonucleotide reductase, the essential enzyme for DNA synthesis, and a potent general protein disulfide reductase with numerous functions in growth and redox regulations. Specific protein disulfide targets for reduction by Trx include protein disulfide-isomerase (PDI) and a number of transcription factors such as p53 , NF-kB and AP-1 (T1-151). Trx is also capable of removing H2O2, particularly when it is coupled with either methionine sulfoxide reductase or several isoforms of peroxiredoxins.
Specifications
Specifications
TRX2
Polyclonal
Unconjugated
TXN2
MT TRX, MTRX, Thioredoxin 2, Thioredoxin, mitochondrial, TRX2, TXN2
Rabbit
Antigen Affinity Chromatography
RUO
25828, 56551, 79462
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P97493, P97615, Q99757
TXN2
TRX2 Fusion Protein Ag3640
150 μL
Primary
Human, Mouse, Rat
Antibody