Learn More
TRPS1 Mouse, anti-Human, Clone: 1B1G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68562-1-IG-150UL
 View more versions of this product
Product Code. 18068975
  • 410.00€ / 150µL
Description
The TRPS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRPS1. This antibody recognizes human antigen. The TRPS1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TRPS1
1B1G2
GC79, LGCR, TRPS1, Zinc finger protein GC79
150 μL
7227
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa