missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35247-20ul
This item is not returnable.
View return policy
Description
TRPC1 Polyclonal antibody specifically detects TRPC1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TRPC1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| HTRP-1, MGC133334, MGC133335, transient receptor potential canonical 1, transient receptor potential cation channel, subfamily C, member 1, transient receptor potential channel 1, Transient receptor protein 1, TRP-1, TRP1short transient receptor potential channel 1, TrpC1 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human TRPC1 (NP_001238774.1).,, Sequence:, LLNLYSLVYNEDKKNTMGPALERIDYLLILWIIGMIWSDIKRLWYEGLEDFLEESRNQLSFVMNSLYLATFALKVVAHNKFHDFADRKDWDAFHPTLVAEG | |
| 20 μL | |
| Cardiovascular Biology, Neuroscience, Signal Transduction | |
| 7220 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction