missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRP7 Antibody (N64A/36), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-48318-0.025mg
This item is not returnable.
View return policy
Description
TRP7 Monoclonal specifically detects TRP7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications
| TRP7 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-10 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 0.1-1.0 μg/mL | |
| hTRP7, KNP3, likley ortholog of mouse transient receptor potential cation channel, subfamilyC, member 7, putative capacitative calcium channel, short transient receptor potential channel 7, transient receptor potential cation channel, subfamily C, member 7, transient receptor potential-related channel 7, a novel putative Ca2+ channelprotein10TRPM2, Transient receptor protein 7, TRP7, TRP-7, TrpC7 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 57113 | |
| Human | |
| Purified |
| Western Blot | |
| S64A-36 | |
| Unconjugated | |
| Q9HCX4, NP_065122.1 | |
| TRPC7 | |
| Synthetic peptide amino acids 845-862 of human TRPC7 | |
| 0.025 mg | |
| Primary | |
| Detects approx 100kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction