missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TROY Monoclonal Antibody (323606)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Tumor necrosis factor receptor superfamily member 19, commonly known as TROY, is a gene located on chromosome 1p36.22 that encodes a member of the tumor necrosis factor receptor (TNFR) superfamily. TROY is expressed in the nervous system, skin, and various tissues, where it plays a role in regulating cell apoptosis, inflammation, and development. As a receptor, TROY interacts with ligands and other signaling molecules to modulate pathways involved in cell survival, differentiation, and immune responses. In the context of the nervous system, TROY has been implicated in axonal outgrowth and regeneration, particularly influencing signaling pathways related to neural development and repair. Additionally, TROY has been studied in cancer research due to its potential role in tumor growth and progression, making it a candidate for therapeutic targeting. Its involvement in diverse biological processes highlights its significance in maintaining cellular homeostasis and its potential as a therapeutic target in neurobiology and oncology.
Specifications
Specifications
| Antigen | TROY |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 323606 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | Tnfrsf19 |
| Gene Accession No. | Q9JLL3 |
| Gene Alias | AL023044; AW123854; RGD1564996; TAJ; TAJ-alpha; TNF receptor superfamily member 19; Tnfrsf19; Toxicity and JNK inducer; TRADE; TROY; tumor necrosis factor receptor superfamily member 19; tumor necrosis factor receptor superfamily, member 19; UNQ1888/PRO4333 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?