Learn More
Troponin C (cardiac) Antibody (OTI5H3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74638AF405
This item is not returnable.
View return policy
Description
Troponin C (cardiac) Monoclonal specifically detects Troponin C (cardiac) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| Troponin C (cardiac) | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TNNC1 | |
| Full length recombinant protein of human Troponin C (cardiac) (NP_003271) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Immunohistochemistry | |
| OTI5H3 | |
| Immunohistochemistry | |
| cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow | |
| Mouse | |
| Affinity Purified | |
| Cellular Markers, Cytoskeleton Markers | |
| 7134.0 | |
| Store at 4°C in the dark. |