Learn More
Melford Trisodium Citrate, Dihydrate, [Citric Acid, Trisodium Dihydrate]
Product Code. 30018779
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018779 5 kg 5kg
30018778 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018779 Supplier Melford Supplier No. S230405000.0
 Restricted Item
Estimated Shipment: 28-10-2026
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
CAS Number: 6132-04-3
Molecular Formula: C6H5Na3O7 2H2O
Molecular Weight: 294.1
Specifications
CAS 4-3-6132
Molecular Formula C6H5Na3O7-2H2O
Quantity 5 kg