missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, >99%, Reagent grade, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Description
Cell Biology and Analysis
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 5 kg |
| Assay Percent Range | >99% |
| Formula Weight | 157.6 |
| Grade | Reagent |
| Chemical Name or Material | Tris Hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction