Learn More
Tris-Glycine-SDS, 10X Powder (Electrophoresis), Fisher BioReagents
Product Code. 10061653
Click to view available options
Quantity:
1 Pk.
Unit Size:
Each
Product Code. 10061653
Brand: Fisher BioReagents BP13421
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Tris-Glycine-SDS is commonly used as a buffer in SDS-PAGE.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tris-Glycine-SDS
Name Note 10X Powder
CAS 77-86-1, 56-40-6, 151-21-3
Color White
Physical Form Solid
Quantity 1 Pk.
Packaging Foil Pack
Protease Protease free
Grade Electrophoresis
compliance-icons
Product Identifier
  • Tris Glycine SDS 10X Powder
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.