missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl-1-octylammonium bromide, 97%

Product Code. 11372606
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11372606

Brand: Thermo Scientific Alfa Aesar L08610.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2083-68-3 | C11H26BrN | 252.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2083-68-3
Molecular Formula C11H26BrN
Molecular Weight (g/mol) 252.24
MDL Number MFCD00051873
InChI Key XCOHAFVJQZPUKF-UHFFFAOYSA-M
Synonym n-octyltrimethylammonium bromide, octalone, octyltrimethylammonium bromide, n,n,n-trimethyloctan-1-aminium bromide, 1-octanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethyl-1-octanaminium bromide, trimethyloctylammonium bromide 6ci,7ci, n-octyltrimethylammoniumbromide, trimethyloctylamine, bromide, trimethyl octyl azanium bromide
PubChem CID 74964
ChEBI CHEBI:346954
IUPAC Name trimethyl(octyl)azanium;bromide
SMILES CCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 220°C to 222°C
Quantity 25 g
Beilstein 3628448
Sensitivity Hygroscopic
Formula Weight 252.24
Percent Purity 97%
Chemical Name or Material Trimethyl-1-octylammonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.