accessibility menu, dialog, popup

UserName

CAS RN 2083-68-3

BrH3CNCH3CH3H3C

CAS RN 2083-68-3

IUPAC Name: trimethyl(octyl)azanium bromide
Synonyms: octalone
Molecular Weight (g/mol): 252.24
Molecular Formula: C11H26BrN
InChi Key: XCOHAFVJQZPUKF-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCC[N+](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results