missing translation for 'onlineSavingsMsg'
Learn More

TRIM72 Mouse Monoclonal Antibody, Proteintech

Product Code. 30237373 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30237373 150 μL 150µL
30234830 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30237373 Supplier Proteintech Supplier No. 674031IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TRIM72 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRIM72. This antibody recognizes human, mouse, pig antigen. The TRIM72 antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.

Specifications

Antigen TRIM72
Applications Western Blot, Flow Cytometry (Intracellular Staining), ELISA
Classification Monoclonal
Clone 5F2C11
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493829
Target Species Human, Mouse, Pig
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 53 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.