missing translation for 'onlineSavingsMsg'
Learn More

TRIM60 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

484.00€

Specifications

Antigen TRIM60
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18267263
View Documents
Novus Biologicals
NBP1-55076
100 μL
484.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TRIM60 Polyclonal specifically detects TRIM60 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TRIM60
Polyclonal
Rabbit
Zinc Finger
Q495X7
166655
Synthetic peptides corresponding to TRIM60(tripartite motif-containing 60) The peptide sequence was selected from the N terminal of TRIM60. Peptide sequence LEGSLEPLRNNIERVEKVIILQGSKSVELKKKVEYKREEINSEFEQIRLF.
Primary
Western Blot
Unconjugated
RUO
Human
FLJ35882, RING finger protein 129MGC119325, RING finger protein 33, RNF129, RNF33, tripartite motif containing 60, tripartite motif-containing 60, tripartite motif-containing protein 60
TRIM60
IgG
55 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.