missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM44 Antibody (9A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43053
This item is not returnable.
View return policy
Description
TRIM44 Monoclonal antibody specifically detects TRIM44 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| TRIM44 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 54765 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 9A2 | |
| Western Blot 1:500 - 1:5000, ELISA, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| DIPBMGC3490, HSA249128, MC7, Protein DIPB, tripartite motif containing 44, tripartite motif-containing 44, tripartite motif-containing protein 44 | |
| tripartite motif-containing 44 - (Uniprot# Q96DX7) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction