missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35620-20ul
This item is not returnable.
View return policy
Description
TRIM4 Polyclonal antibody specifically detects TRIM4 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TRIM4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| RING finger protein 87, RNF87tripartite motif protein 4, tripartite motif containing 4, tripartite motif-containing 4, tripartite motif-containing protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TRIM4 (NP_148977.2).,, Sequence:, MEAEDIQEELTCPICLDYFQDPVSIECGHNFCRGCLHRNWAPGGGPFPCPECRHPSAPAALRPNWALARLTEKTQRRRLGPVPPGLCGRHWEPLRLFCEDDQRPVCLVCRESQEHQTHAMAPIDEAFESYREKLLKSQRNLVAKMKKVMHLQDVEVKNATQWKDKIKSQRMRISTEFSKLHNFLVEEEDLFLQRLNKEEEETKKKLNENTLKLNQTIASLKKLILEVGEKSQAPTLELLQNPKEVLTRSEIQDVNYSLEAVKVKTVCQIPLMKEMLKRFQ | |
| 20 μL | |
| Zinc Finger | |
| 89122 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction