missing translation for 'onlineSavingsMsg'
Learn More

TRIM38 Monoclonal Antibody (OTI15H4), TrueMAB™, OriGene

Product Code. 17585172
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17585172 30 μL 30µL
17565172 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17585172 Supplier Origene Technologies Supplier No. TA505976S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRIM38 Monoclonal antibody specifically detects TRIM38 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen TRIM38
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI15H4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CTAG1B
Gene Accession No. P78358
Gene Alias E3 ubiquitin-protein ligase TRIM38; Gm23; hypothetical protein LOC531022; MGC8946; RING finger protein 15; RING-type E3 ubiquitin transferase TRIM38; RNF15; Ro/SSA ribonucleoprotein homolog; RORET; TRIM38; tripartite motif containing 38; tripartite motif-containing 38; tripartite motif-containing protein 38; tripartite motif-containing protein 38; E3 ubiquitin-protein ligase TRIM38; tripartite motif-containing protein 38; LOW QUALITY PROTEIN: E3 ubiquitin-protein ligase TRIM38; zinc finger protein RoRet
Host Species Mouse
Immunogen Full length human recombinant protein of human CTAG1B produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1485
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.