Learn More
Trifluoroacetic acid, ammonium salt, 98%
Product Code. 10627022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627022 25 g Glass bottle 25g
10083573 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10627022 Supplier Thermo Scientific Acros Supplier No. 218860250
  • ONLINE EXCLUSIVE PRICE
    56.05€
    51.30€ / 25g
    Save 4.75€
    8% Off
 Restricted Item
In Stock
Add to basket
CAS: 3336-58-1 | C2H5F3NO2 | 132.06 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3336-58-1
Molecular Formula C2H5F3NO2
Molecular Weight (g/mol) 132.06
MDL Number MFCD00012615,MFCD03095537
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonym ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
PubChem CID 86601334
IUPAC Name azanium;2,2,2-trifluoroacetic acid
SMILES [NH4+].OC(=O)C(F)(F)F
Specifications
Melting Point 122.0°C to 126.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Glass bottle
Linear Formula CF3CO2NH4
Quantity 25 g
Beilstein 02,II,186
Solubility Information Solubility in water: 100g/L (25°C)
Formula Weight 131.05
Percent Purity 0.98
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Trifluoroacetic acid, ammonium salt, 0.98
Show More Show Less
compliance-icons
Product Identifier
  • Trifluoroacetic acid, ammonium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.